Learn More
Tocris Bioscience™ VU 0483605 Available on GSA/VA Contract for Federal Government customers only.
Positive allosteric modulator of mGlu1 receptors
$162.10 - $658.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
53-771-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5377/10
Each for $162.10
Add to cart
53-775-0 View Documents Tocris Bioscience™
5377/50
Each for $658.50
Add to cart
Description
Description
Positive allosteric modulator at mGlu1 (EC50 values are 356 and 390 nM for rat and human receptors, respectively). Exhibits >25-fold selectivity for mGlu1 over mGlu4. Potentiates the decreased glutamate-induced calcium response in mGlu1 mutant cell models of schizophrenia.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.