Learn More
VU 29, Tocris™
Potent mGlu5 allosteric potentiator
Manufacturer:  R&D Systems 4458/10
Quantity 10mg
 View more versions of this product
Catalog No. 44-581-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
384.39
VU 29
C22H16N4O3
N-(1, 3-Diphenyl-1H-pyrazolo-5-yl)-4-nitrobenzamide
Specifications
10mg
>99%
384.39
VU 29
890764-36-0
C22H16N4O3
N-(1, 3-Diphenyl-1H-pyrazolo-5-yl)-4-nitrobenzamide
Safety and Handling
Recommended Storage : Store at Room Temperature