Learn More
VU 591 hydrochloride, Tocris™
Selective Kir 1.1 (ROMK) inhibitor
Manufacturer:  R&D Systems 4197/50
Quantity 50mg
 View more versions of this product
Catalog No. 41-975-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
368.3
VU 591 hydrochloride
C16H12N6O5
6-Nitro-2-[(6-nitro-1H-benzimidazol-2-yl)methoxymethyl]-1H-benzimidazole hydrochloride
Specifications
50mg
>99%
368.3
VU 591 hydrochloride
1315380-70-1
C16H12N6O5
6-Nitro-2-[(6-nitro-1H-benzimidazol-2-yl)methoxymethyl]-1H-benzimidazole hydrochloride
Safety and Handling
Recommended Storage : Desiccate at Room Temperature