Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VUF 10166, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity 5-HT3 receptor antagonist (IC50 values are 0.04 and 22 nM for human 5HT3A and 5-HT3AB receptors respectively). Exhibits partial agonist activity at 5-HT3A receptors at higher concentrations (EC50 = 5.2 μM). Also histamine H4 receptor antagonist (pKi = 6.64 in HEK cells).
Specifications
Quantity | 10 mg |
Formula Weight | 262.74 |
Percent Purity | >98% |
Chemical Name or Material | VUF 10166 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction