Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VUF 5681 dihydrobromide, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
639089-06-8
C11H21Br2N3
355.118
YFFVIGXPKWSZHW-UHFFFAOYSA-N
vuf 5681 dihydrobromide,4-3-1h-imidazol-4-yl propyl piperidine dihydrobromide,4-3-1h-imidazol-5-yl propyl piperidine-hydrogen bromide 1/2,4-3-3h-imidazol-4-yl propyl piperidine dihydrobromide
53440140
4-[3-(1H-imidazol-5-yl)propyl]piperidine;dihydrobromide
C1CNCCC1CCCC2=CN=CN2.Br.Br
This item is not returnable. View return policy
639089-06-8
C11H21Br2N3
355.118
YFFVIGXPKWSZHW-UHFFFAOYSA-N
vuf 5681 dihydrobromide,4-3-1h-imidazol-4-yl propyl piperidine dihydrobromide,4-3-1h-imidazol-5-yl propyl piperidine-hydrogen bromide 1/2,4-3-3h-imidazol-4-yl propyl piperidine dihydrobromide
53440140
4-[3-(1H-imidazol-5-yl)propyl]piperidine;dihydrobromide
C1CNCCC1CCCC2=CN=CN2.Br.Br

Potent H3 receptor silent antagonist

Potent histamine H 3 receptor silent antagonist (pK i = 8.35).
Quantity 50mg
Formula Weight 355.11
Percent Purity >98%
Chemical Name or Material VUF 5681 dihydrobromide

Recommended Storage : Desiccate at Room Temperature