VUF 5681 dihydrobromide, Tocris

Potent H3 receptor silent antagonist

Manufacturer: R&D Systems 2493/10

Quantity 10mg

 View more versions of this product

Catalog No. 24-931-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C11H19N3·2HBr
    Formula Weight 355.11
    Synonym 4-[3-(1H-Imidazol-4-yl)propyl]piperidine dihydrobromide
    Chemical Name or Material VUF 5681 dihydrobromide

    Specifications

    Formula Weight 355.11
    Quantity 10mg
    CAS 639089-06-8
    Synonym 4-[3-(1H-Imidazol-4-yl)propyl]piperidine dihydrobromide
    Assay Percent Range >98%
    Molecular Formula C11H19N3·2HBr
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material VUF 5681 dihydrobromide

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit