Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Cole Parmer Neytech 9495130 Vulcan Muffle Furnace, 565 cu in, Digital, Single Set Point; 120 VAC DFS Item
Manufacturer:  Cole Parmer 3385526
Combined fiber and firebrick insulation cools quickly and provides extended heating element life. Heating elements slide out for quick replacement. Vertical lift door directs heat away from user and saves counter space. Suitable for wax burnout, ashing, heat treating, ceramic and glass seal firing.
Catalog No. NC1469941