Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWC2 Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA548096
Description
In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Brorin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Specifications
VWC2 | |
Polyclonal | |
Unconjugated | |
Vwc2 | |
A930041G11Rik; brain-specific chordin-like protein; Brorin; CR (chordin-like cysteine-rich) domain-containing adhesive protein; Cradin; G11; PSST739; UNQ739; UNQ739/PRO1434; von Willebrand factor C domain containing 2; von Willebrand factor C domain-containing protein 2; Vwc2 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
375567 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q2TAL6 | |
Vwc2 | |
Mouse myeloma cell line NS0-derived recombinant human Brorin/VWC2 Ser28-Met325. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction