Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VWCE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551338
Description
The antibody detects endogenous levels of total VWCE protein.
May be a regulatory element in the beta-catenin signaling pathway and a target for chemoprevention of hapatocellular carcinoma.
Specifications
VWCE | |
Polyclonal | |
Unconjugated | |
VWCE | |
1300015B04Rik; HBV X protein up-regulated gene 11 protein; HBxAg up-regulated gene 11 protein; RGD1309910; URG11; von Willebrand factor C and EGF domain-containing protein; von Willebrand factor C and EGF domains; VWC1; Vwce | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220001 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96DN2 | |
VWCE | |
Synthetic peptide of human VWCE. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction