Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/1465) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$627.50
Specifications
Antigen | vWF-A2 |
---|---|
Clone | VWF/1465 |
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug protein lysate, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254430100
|
Novus Biologicals
NBP254430100UG |
100 μg |
Each for $627.50
|
|
|||||
NB254430200
|
Novus Biologicals
NBP254430200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.Specifications
vWF-A2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
F8VWF, VWD, VWF von Willebrand factor | |
VWF | |
IgG1 | |
Protein A or G purified | |
250 kDa |
VWF/1465 | |
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug/500ug protein lysate, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with No Preservative | |
7450 | |
Recombinant fragment of human vWF protein (aa1815-1939) (exact sequence is proprietary) | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title