Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Mac Mod AnalyticalSupplier Diversity Partner VYD. 214MS 5 C4 50X4.6MM DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 214MS5405
Vydac 214MS 5um C4 Column 50x4.6mm Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 214MS5405 Alternative Part Number: 5104305
Catalog No. 50-112-0568