Learn More
Mac Mod AnalyticalSupplier Diversity Partner VYD. 214TP 5 C4 150X2.1MM DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 214TP5215
Vydac 214TP 5um C4 Column 150x2.1mm Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 214TP5215 Alternative Part Number: 5103924
Catalog No. 50-112-0840