Learn More
Mac Mod AnalyticalSupplier Diversity Partner VYD 218MS 10 C18 GRD 3-4.6MM DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 218GD104MSN
Vydac 218MS 10um C18 Guard Cartridge (for 3.0 - 4.6mm id columns) 2pk Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 218GD104MS Alternative Part Number: 5104145
Catalog No. 50-112-0113