Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Mac Mod AnalyticalSupplier Diversity Partner VYD. 218TP 10 C18 250X22MM DFS Item
Manufacturer:  Mac Mod AnalyticalSupplier Diversity Partner 218TP1022
Vydac 218TP 10um C18 Column 250x22mm Note: All Grace Analytical and Semi-Preparative HPLC Columns brands including Vydac®, Alltima®, Alltima® HP, Prevail™, Apollo™, Allsep®, Apex™ and Genesis™ analytical HPLC column ranges are now available in the US from MAC-MOD Analytical via Fisher Scientific - Hichrom Limited recently obtained worldwide manufacturing rights to these brands and is offering them in the US through MAC-MOD Analytical. These columns are made with the same exact manufacturing protocols and to the identical specifications previously used by Grace. Category: Chromatography Columns and Supplies GRACE Part Number: 218TP1022 Alternative Part Number: 5107107
Catalog No. 50-112-0215