Learn More
Thermo Scientific™ Vytac™ Formalin Disposal System Available on GSA/VA Contract for Federal Government customers only.
Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Thermo Scientific™ Vytac™ Formalin Disposal System.
$148.00 - $554.50
Products 2
Catalog Number Mfr. No. Item Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Item Description Quantity Price Quantity & Availability  
22-046-332 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
5401
Vytac; 1 Gal. (3.79L) 1 Gal. (3.79L) Each for $148.00
Case of 4 EA for $498.26
Add to cart
22-046-360
View Documents
Thermo Scientific™
5405
Vytac; 5 Gal. (18.9L) 5 Gal. (18.9L) Each for $554.50
Add to cart
Description
Description
Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Thermo Scientific™ Vytac™ Formalin Disposal System. This simple, economical system eliminates the need for expensive waste hauling and onsite permits. 
Disposal Solution:
  • Breaks down formalin into inert components
  • Neutralizes four parts formalin waste per one part Vytac
  • When used as directed, neutralizes more than 99% of the waste formalin collected
Note: Vytac GNF will neutralize gluteraldehyde
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.