Learn More
Tocris Bioscience™ W-84 dibromide
Antagonist-receptor complex stabilizer
Supplier: Tocris Bioscience™ 0532/10
This item is not returnable.
View return policy
Description
Stabilizes cholinergic antagonist-receptor complexes by an allosteric effect. Increases the protective effect of atropine against organophosphate poisoning.Specifications
Hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide | |
10 mg | |
C32H44Br2N4O4 |
21093-51-6 | |
Soluble to 10mM in DMSO | |
Observed MW: 708.53 |
Safety and Handling
Recommended Storage : Store at Room Temperature