Learn More
Tocris Bioscience™ W-84 dibromide
Antagonist-receptor complex stabilizer
Supplier:  Tocris Bioscience™ 0532/50
 View more versions of this product
Catalog No. 05-325-0
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
Stabilizes cholinergic antagonist-receptor complexes by an allosteric effect. Increases the protective effect of atropine against organophosphate poisoning.
Specifications
Specifications
Hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide
50 mg
C32H44Br2N4O4
21093-51-6
Soluble to 10mM in DMSO
Observed MW: 708.53
Safety and Handling
Safety and Handling
Recommended Storage : Store at Room Temperature