Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Tocris Bioscience™ W-84 dibromide

$145.00 - $609.00

Specifications

Formula Weight Observed MW: 708.53
Chemical Name or Material Hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide
Molecular Formula C32H44Br2N4O4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05-325-0 View Documents Tocris Bioscience™
0532/50
50mg Each for $609.00
N/A
05-321-0 View Documents Tocris Bioscience™
0532/10
10mg Each for $145.00
N/A
Description

Description

Stabilizes cholinergic antagonist-receptor complexes by an allosteric effect. Increases the protective effect of atropine against organophosphate poisoning.
Specifications

Specifications

Observed MW: 708.53
Hexamethylene-bis-[dimethyl-(3-phthalimidopropyl)ammonium]dibromide
C32H44Br2N4O4
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit