Learn More
Sklar InstrumentsSupplier Diversity Partner Wachenfeldt Wound Clips, Sklar, 16mm, Pointed, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 241116
Wachenfeldt Wound Clips, Sklar, 16mm, Pointed, Stainless Steel. Sklar's Wachenfeldt Clips may be applied to wounds and removed when healing is complete. The are also used temporarily as part of a procedure to stop blood flow. The clips are available in a variety of sizes to accommodate different incisions.
Catalog No. 50-143-8001