Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wagner Milwaukee™ Heat Guns

Rugged industrial heat guns provide up to 5700 BTUs and 1680w of power

Manufacturer:  Wagner 10018

 View more versions of this product

Catalog No. 09-201-13

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Adjustable hot-air temperature up to 540°C (1000°F); also cool-air setting
  • Body is diecast magnesium; heating element is replaceable
  • With stationary stand for hands-free operation

Order Info

For 120V 50/60Hz

Certifications

UL listed

Specifications

Specifications

14A
1680 W
120V, 50/60Hz
50/60 Hz
Heat Gun
120V
UL
Stationary stand for hands-free operation, cool air setting
400° to 540°C
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit