Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversified Spaces™ Wall Cabinet with Solid Double Doors
SCI_ED

Cabinets can be ganged together to create a wall of base cabinets.

Supplier:  Diversified Spaces™ D033612

 View more versions of this product

Catalog No. S47295



Description

Description

  • Has 2 adjustable shelves
  • Constructed of solid oak and oak veneers
  • Natural with chemical resistant UV finish
  • Hanging wall cabinet with locking double doors

Compliance

SEFA

Specifications

Specifications

MAS Certified Green
30.4 cm
Natural with chemical resistant UV
76.2 cm
Limited Lifetime
91.4 cm
12 in.
Solid double doors
30 in.
Red Oak
36 in.
Wall Mounted Cabinet
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit