Learn More
Detecto Adult Stadiometer, Mechanical, Height range: 30 to 78 inch (76 to 200 cm), 1/4 inch (1/2 cm) graduations. Shipping weight: 8 lb / 3.6 kg.  
Supplier:  Detecto 3PHTRODWM
Encompass_Preferred
Detecto’s stadiometers have long been the choice for accurate medical height measurement. Everything needed for operation is included with the stadiometer and installation takes seconds with the included hardware and built-in wall-mount bracket. Merely lift the spoon and request for the patient to step up to be measured. Easy to install, easy to operate, and with durable steel construction for longevity and reliability. Height range: 30 to 78 inch (76 to 200 cm), 1/4 inch (1/2 cm) graduations. Shipping weight: 8 lb / 3.6 kg.
Catalog No. NC0407658