Learn More
Contec™ Wall WipR™ Cleaning System and Accessories
Offers wide surface coverage and low operator fatigue
Manufacturer:  Contec™ WWH18
 View more versions of this product
Catalog No. 19-038-563
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Polyester fabric combined with a lightweight aluminum frame and handle offers wide surface coverage and low operator fatigue
  • The specially-designed fabric covers are low in extractable residues and even lower in particle generation
  • Long, lightweight handle allows easy cleaning of ceilings and walls
  • Swivel “T” handle, 18 in. (45.7cm)
  • Polyester/foam wiping cover, 18 in. (45.7cm)
  • Aluminum extension handle, 8 ft. or 14 in.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Specifications
Specifications
Aluminum, Aluminum
T-Handle
Contec™ Cleaning System
T Handle
Cleaning system is designed to clean walls and ceilings in controlled environments