Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ Wallerstein Broth (WL) 2mL Ampoules (50 Pack)

Ready-to-use 2mL ampoule Wallerstein Broth (WL) is a liquid culture growth media that can save time and money by shortening prep time and minimizing possible contamination. The 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish

$102.92 - $333.00

Specifications

Manufacturing Origin USA
Packaging Ampoules
Packaging Quantity 2 mL
Sterility Sterile
View More Specs

Products 2
Catalog Number Mfr. No. Broth Type Detects Price Quantity & Availability  
Catalog Number Mfr. No. Broth Type Detects Price Quantity & Availability  
10-486-143 View Documents GVS™
10496108
Wallerstein Nutrient Broth (WL) for cultivation and enumeration of yeast Pack of 50 for $333.00
Add to cart
 
10-486-144 View Documents GVS™
10496109
Wallerstein Differential Broth (WLD) For determination of bacterial count. Pack of 50 for $102.92
Add to cart
 
Description

Description

Description

  • WL Nutrient Broth is for the cultivation and enumeration of yeast and WL Differential Broth is for determination of bacterial count. Use of the medium at pH 5.5 and incubation at 25°C will give reliable counts for brewer’s yeast. Adjustment of the pH to 6.5 and incubation at 30°C allows for the selective growth of baker’s and distiller’s yeast

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 2 mL Portions, Reduce Risk of Cross-Contamination.
  • 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • Use with membrane filters to culture bacteria, yeasts, and molds.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits. This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Specifications

USA
Ampoules
2 mL
Sterile
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit