Learn More
Walter Products Digital Water Baths
Features high precision PID temperature controls.
Supplier: Walter Products HW4L
Description
- LCD display allows user to select and monitor temperature setting
- Over temperature alarm provides reliable protection for samples
- Steel exterior with electrostatic coating
- Stainless-steel inner chamber, cover and concentric rings for corrosion resistance and durability
Specifications
Water Bath w/Two Cover Openings | |
5°C to 99°C | |
Stainless Steel | |
11.7 cm | |
12 kg | |
500 w |
4 L | |
LCD Digital | |
12 x 6 x 8 in. | |
26.5 lb. | |
110 V | |
50/60 Hz |