Learn More
Sklar InstrumentsSupplier Diversity Partner Walther Dilator-Catheter, Sklar, 5-1/2 in., Curved, Chrome Plated Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 852024
Walther Dilator-Catheter, Sklar, 5-1/2 in., Curved, Chrome Plated Steel. Sklar's Walther Dilator-Catheter is a female urethral sound commonly used to dilate the female urethra before placement of a cystoscope to ease insertion. It may also be used to take a urine specimen or drain the bladder. This product has a 24 French (8mm) curved tip and overall length of 5-1/4 inches.
Catalog No. 50-143-6405