Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wang resin, 1% crossl with DVB, 0.5-0.7mmol/g, 200-400 mesh, ACROS Organics™

Click to view available options
Quantity:
25g
201058-08-4
C15H16O2
228.291
MFCD00131778
NERFNHBZJXXFGY-UHFFFAOYSA-N
[4-(Hydroxymethyl)phenoxymethyl]polystyrene divinylbenzene copolymer
2734007
[4-[(4-methylphenyl)methoxy]phenyl]methanol
CC1=CC=C(C=C1)COC2=CC=C(C=C2)CO
201058-08-4
C15H16O2
228.291
MFCD00131778
NERFNHBZJXXFGY-UHFFFAOYSA-N
[4-(Hydroxymethyl)phenoxymethyl]polystyrene divinylbenzene copolymer
2734007
[4-[(4-methylphenyl)methoxy]phenyl]methanol
CC1=CC=C(C=C1)COC2=CC=C(C=C2)CO
for solid support synthesis
Additional Information crosslinking: 1% DVB
Loading 0.5 to 0.7mmol/g
Mesh Size 200 to 400 mesh
Quantity 25g
Color Beige
Physical Form Beads
Chemical Name or Material Wang resin