Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Conair™ Waring™ Laboratory Blenders: Two Speeds, Commercial
Commercial-grade units for demanding mixing and blending applications
Supplier: Conair™ 8011S
Includes: Base, container, 3-wire grounded cord and plug
Description
Commercial-grade units for demanding mixing and blending applications
- Cloverleaf design
- Choice of glass and/or stainless-steel containers
- Sturdy, diecast metal bases and stainless-steel blades
- All durably constructed for years of dependable service
- Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
- Heat-resistant borosilicate glass (integral handle) or stainless-steel (attached handles)
- Two-piece vinyl and styrene lids on all models
Order Info
Electrical requirements for all models: 115/120V 50/60Hz
Certifications
UL listed (Select models only; see ordering chart)
Specifications
3.6 A | |
240 V | |
50/60 Hz | |
1000 mL | |
9.75 x 8 x 15.25 in. (24.76 x 20.32 x 39 cm) | |
39 cm | |
24.76 cm | |
20.32 cm | |
Stainless Steel | |
1 Ea. |
18,000, 22,000 rpm | |
240 V, 50/60 Hz | |
0.4 hp | |
34 oz. | |
15.25 in. | |
9.75 in. | |
8 in. | |
Epoxy-coated Metal | |
Base, container, 3-wire grounded cord and plug |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction