Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conair™ Waring™ Laboratory Blenders: Two Speeds, Explosion-Resistant

Extra protection for mising volatile substances

Manufacturer:  Conair™ 707SB

Catalog No. 14-509-7A


Add to cart

Includes: Base, container, 3-wire grounded cord and plug

Description

Description

Extra protection for mising volatile substances

  • Cloverleaf design
  • Explosion-resistant
  • Choice of glass and/or stainless-steel containers
  • Sturdy, diecast metal bases and stainless-steel blades
  • All durably constructed for years of dependable service
  • Epoxy-coated bases with chrome top pieces on all models, except for the aluminum and steel base (Hazardous-Location Model)
  • Heat-resistant borosilicate glass (integral handle)
  • Two-piece vinyl and styrene lid)
x

Order Info

Electrical requirements for all models: 115/120V 50/60Hz

Specifications

Specifications

16 in.
9.62 in.
Each
20.32cm
3.6A
50/60Hz
0.7hp
18000, 22000rpm
34 oz.
115/120V 50/60Hz
41cm
24.43cm
8 in.
Epoxy Coated Die Cast Zinc
Glass
1000mL
115/120V
Base, container, 3-wire grounded cord and plug
9.62 x 8 x 16 in. (24.43 x 20.32 x 41cm)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit