Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Conair™ Waring™ Seven-Speed Blenders

3,500 to 22,000 RPM in 7 steps

Manufacturer:  Conair™ 7012G

 View more versions of this product

Catalog No. 14-509-7G

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

3,500 to 22,000 RPM in 7 steps

Specifications

Specifications

13.87 in.
9.75 in.
Each
20.32cm
7A
50/60Hz
0.7hp
3500 to 22000rpm
33.8 oz.
120VAC 50/60Hz
35.22cm
24.76cm
8 in.
Epoxy Coated Die Cast Zinc
Glass
1000mL
120VAC
Handle and two piece vinyl and styrene lid, stainless steel blade
9.75 x 8 x 13.87 in. (24.76 x 20.32 x 35.22cm)
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit