Learn More
Tip Temperature Products WarmMark TTI Indicator -18C/0F
SDP
Supplier:  Tip Temperature Products WMSSEN001/5099
Encompass
This item has a minimum qty of 35 per supplier requirements.
WarmMark TTI Indicator -18C/0FRising Temp Monitor Changes Color PermanentlyBrief 1 hour, Moderate 3 Hrs, Prolonged 12 Hrs.Field-armable with pull tab**Per supplier requirements this item has a minimum qty of 50.**
Catalog No. NC1496789
Please call customer service for current pricing. \n1-800-766-7000