Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Warning Labels, Pictogram: Dangerous Electrical Voltage
Feature pictograms that indicate dangerous electrical voltage. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.
Supplier: Brady™ 60235
Description
- Made with highly visible B-946 bright colored vinyl
- Adheres to any clean, dry surface
- Available in 5 different sizes
Specifications
Pictogram: Dangerous Electrical Voltage | |
Black, Yellow | |
Warning Label | |
Triangle | |
0.004 in. |
Acrylic, Permanent, Cold Temperature | |
Pressure-sensitive Vinyl (B-946) | |
Indoor/Outdoor | |
0.5 x 0.5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction