Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Warning Labels, Pictogram: Hazardous Area

Feature pictograms that indicate hazardous area. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.

Manufacturer:  Brady™ 60188

 View more versions of this product

Catalog No. 19-107-257


  Check Availability
Add to cart

Specifications

Description

  • Made with highly visible B-946 bright colored vinyl
  • Adheres to any clean, dry surface
  • Available in 5 different sizes
Description & Specifications

Specifications

Acrylic,Cold temperature,Permanent
Pictogram: Hazardous Area
Triangle
0.004 in.
Warning Label
Black,Yellow
Indoor/Outdoor
W x H: 0.5 x 0.5 in.
Pressure Sensitive Vinyl (B-946)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit