Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Warning Labels, Pictogram: Hazardous Area
Feature pictograms that indicate hazardous area. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.
$129.00 - $262.00
Specifications
Description | Pictogram: Hazardous Area |
---|---|
Adhesive Type | Acrylic, Permanent, Cold Temperature |
Color | Black, Yellow |
Material | Pressure-sensitive Vinyl (B-946) |
Product Type | Warning Label |
Catalog Number | Mfr. No. | Size (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size (English) | Price | Quantity & Availability | |||||
19-107-257
|
Brady™
60188 |
0.5 x 0.5 in. |
Pack of 10 for $129.00
|
|
|||||
19-107-264
|
Brady™
60199 |
1 x 1 in. |
Pack of 10 for $131.00
|
|
|||||
19-107-271
|
Brady™
60210 |
2 x 2 in. |
Pack of 10 for $227.00
|
|
|||||
19-107-278
|
Brady™
60221 |
4 x 4 in. |
Pack of 10 for $158.00
|
|
|||||
19-107-285
|
Brady™
60232 |
8 x 8 in. |
Pack of 10 for $262.00
|
|
|||||
Description
- Made with highly visible B-946 bright colored vinyl
- Adheres to any clean, dry surface
- Available in 5 different sizes
Specifications
Pictogram: Hazardous Area | |
Black, Yellow | |
Warning Label | |
Triangle |
Acrylic, Permanent, Cold Temperature | |
Pressure-sensitive Vinyl (B-946) | |
Indoor/Outdoor | |
0.004 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title