Brady™ Warning Labels, Pictogram: Hazardous Area

Feature pictograms that indicate hazardous area. Brady™ Warning Labels will adhere to any clean, dry surface and are available in a variety of sizes.

Manufacturer: Brady™ 60221

 View more versions of this product

Catalog No. 19-107-278

  Check Availability
Add to Cart

    Specifications

    Description Pictogram: Hazardous Area
    For Use With (Application) Indoor/Outdoor
    Label Size (English) W x H: 4 x 4 in.
    Material Pressure Sensitive Vinyl (B-946)
    Adhesive Type Cold Temperature Permanent Acrylic
    Product Type Warning Label
    Shape Triangle
    Color Black on Yellow
    Thickness (English) 0.004 in.

    • Made with highly visible B-946 bright colored vinyl
    • Adheres to any clean, dry surface
    • Available in 5 different sizes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit