Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Warning Sign: ASBESTOS. MAY CAUSE CANCER AND LUNG DAMAGE. AUTHORIZED PERSONNEL ONLY.

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$13.81 - $33.38

Specifications

Color Black/Red/Yellow on White
Message ASBESTOS. MAY CAUSE CANCER AND LUNG DAMAGE. AUTHORIZED PERSONNEL ONLY.
Quantity 1 Ea.
Shape Rectangular
Thickness 0.035 in.
View More Specs

Products 2
Catalog Number Mfr. No. Material Temperature Range (English) Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Material Temperature Range (English) Dimensions (L x W) Price Quantity & Availability  
19-106-366
View Documents
Brady™
144847
Aluminum -40°F to 230°F 14 x 10 in.
Each for $33.38
Add to cart
 
19-106-367
View Documents
Brady™
144850
Polystyrene -40°F to 176°F 10 x 7 in.
Each for $13.81
Add to cart
 
Description

Description

  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
Specifications

Specifications

Black/Red/Yellow on White
1 Ea.
0.035 in.
Aluminum
ASBESTOS. MAY CAUSE CANCER AND LUNG DAMAGE. AUTHORIZED PERSONNEL ONLY.
Rectangular
Non-adhesive
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit