Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Warning Sign: CAUTION - PROPANE

Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

Specifications

Color Black on yellow
Message CAUTION - PROPANE
Quantity 1/Ea.
Mounting Type Corner holes
Width (English) 17 in.
View More Specs

Products 2
Catalog Number Mfr. No. Material Min. Temperature (English) Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Material Min. Temperature (English) Size (English) Price Quantity & Availability  
19-105-286 View Documents Brady™
125703
Aluminum -40°F 17 in. x 7 in. x 0.035 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-105-287 View Documents Brady™
125704
PS (Polystyrene) -40°F 17 in. x 7 in. x 0.0591 in. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications

Specifications

Black on yellow
1/Ea.
17 in.
Rectangle
CAUTION - PROPANE
Corner holes
7 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit