Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Warning Sign: COMBUSTIBLE DIESEL

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$11.25 - $25.30

Specifications

Color Black/red on white
Message COMBUSTIBLE DIESEL
Min. Temperature (English) -40°F
Quantity 1/Ea.
Shape Rectangle
View More Specs

Products 2
Catalog Number Mfr. No. Material Mounting Type Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Material Mounting Type Size (English) Price Quantity & Availability  
19-106-964 View Documents Brady™
37790
Polyester Self-adhesive 10 in. x 7 in. x 0.01 in. Each for $11.25
Add to cart
 
19-106-975 View Documents Brady™
37833
Aluminum Corner holes 14 in. x 10 in. x 0.035 in. Each for $25.30
Add to cart
 
Description

Description

  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor and outdoor use
Specifications

Specifications

Black/red on white
-40°F
Rectangle
COMBUSTIBLE DIESEL
1/Ea.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit