Learn More
Aviva Systems Biology Corp WARS Antibody - N-terminal region (ARP58550_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58550_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against WARS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human WARS. Alternate names: GAMMA-2, IFI53, IFP53ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62353