Learn More
WASL Mouse anti-Human, Mouse, Clone: OTI9F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811284

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The Wiskott-Aldrich syndrome (WAS) family of proteins share similar domain structure, and are involved in transduction of signals from receptors on the cell surface to the actin cytoskeleton. The presence of a number of different motifs suggests that they are regulated by a number of different stimuli, and interact with multiple proteins. Recent studies have demonstrated that these proteins, directly or indirectly, associate with the small GTPase, Cdc42, known to regulate formation of actin filaments, and the cytoskeletal organizing complex, Arp2/3. The WASL gene product is a homolog of WAS protein, however, unlike the latter, it is ubiquitously expressed and shows highest expression in neural tissues. It has been shown to bind Cdc42 directly, and induce formation of long actin microspikes.Specifications
WASL | |
Monoclonal | |
Unconjugated | |
WASL | |
N-WASP, NWASP, WASPB | |
Mouse | |
Affinity Chromatography | |
RUO | |
73178, 8976 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI9F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00401, Q91YD9 | |
WASL | |
Full length human recombit protein of human WASL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.