Learn More
PicoSolutions WASP Clay Tanks
SCI_ED
WASP Clay Tanks are for the LDM extrusion system
$263.00 - $421.00
Specifications
For Use With (Equipment) STEM Project
View More Specs
Products 3
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S81227
SCI_ED
View Documents
PicoSolutions
PS7035
0.5 L Clay Tank
Each for $263.00
Add to cart
S81225
SCI_ED
View Documents
PicoSolutions
PS7033
3 L Clay Tank
Each for $315.50
Add to cart
S81226
SCI_ED
View Documents
PicoSolutions
PS7034
5 L Clay Tank
Each for $421.00
Add to cart
Description
Description
  • Ideal for ceramic materials
  • Compatible with Delta WASP 2040, 4070, 2040 Clay, 40100 Clay
  • Certified Aluminum up to 40 bar
  • Safety Valve set at 7 bar
Specifications
Specifications
STEM Project