American Educational Products Watch Glass Form Covers

Manufacturer: American Educational Products 702610H

 View more versions of this product

Catalog No. 02-610H

  Check Availability
Add to Cart

    Specifications

    Diameter (English) 6 in.
    Diameter (Metric) 15.24cm
    For Use With (Application) Used for covering boiling solutions
    For Use With (Equipment) For 30 to 4000mL low form beaker, also for funnels, dishes, and jars
    Shape Concave

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit