Learn More
American Educational Products Watch Glass Form Covers
Manufacturer:  American Educational Products 702610J
 View more versions of this product
Catalog No. 02-610J
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Concave shape prevents sliding. Returns condensation to beaker. Great for covering boiling solutions. For 30 to 4000mL low-form beakers. Also for funnels, dishes, and jars. Fire-polished edges for strength.
Specifications
Specifications
8 in.
Used for covering boiling solutions
Concave
20.32cm
For 30 to 4000mL low form beaker, also for funnels, dishes, and jars