Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
American Educational Products Watch Glass Form Covers
Manufacturer: American Educational Products 702610C
Description
Concave shape prevents sliding. Returns condensation to beaker. Great for covering boiling solutions. For 30 to 4000mL low-form beakers. Also for funnels, dishes, and jars. Fire-polished edges for strength.
Specifications
3 in. | |
Used for covering boiling solutions | |
Concave |
7.62cm | |
For 30 to 4000mL low form beaker, also for funnels, dishes, and jars |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title