Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with 0.05% Acetic Acid, LC-MS Ultra, Honeywell™ Riedel-de-Haën™ CHROMASOLV™

Catalog Number 60044852
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
64-19-7, 7732-18-5
H2O
64-19-7, 7732-18-5
H2O

CHROMASOLV™ LC-MS Ultra, LabReady™ Solvent Blend, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

 

TRUSTED_SUSTAINABILITY
UN Number NONH for all modes of transport
Formula Weight 18.02g/mol
Grade LC-MS Ultra CHROMASOLV™
Quantity 2 L
Packaging Glass Bottle
Chemical Name or Material Water With 0.05% Acetic Acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit