Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, 0.05% Trifluoroacetic Acid, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O
This item is not returnable. View return policy
7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
962
CHEBI:15377
oxidane
O

Function tested and optimized for LC/MS applications

    For use in liquid chromatography and mass spectrometry; For laboratory, research, or manufacturing use
Grade BAKER ANALYZED™ LC-MS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Water, 0.05% Trifluoroacetic Acid