Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, 0.1% Trifluoroacetic Acid, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

7732-18-5
H2O
18.015
XLYOFNOQVPJJNP-UHFFFAOYSA-N
962
CHEBI:15377

Function tested and optimized for LC/MS applications

    For use in liquid chromatography and mass spectrometry; For laboratory, research, or manufacturing use
CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Water, 0.1% Trifluoroacetic Acid
Grade BAKER ANALYZED™ LC-MS Reagent
Show More Show Less