Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water with 0.1% trifluoroacetic acid, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS, LabReady™ Solvent Blend
Supplier: Honeywell Chemicals 349782.5L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
For Use With: LC-MS, Contains 0.093-0.107 % (w/w) trifluoroacetic acid as additive | |
2.5 L | |
LC-MS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction