Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water with 0.1% (v/v) Formic Acid, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™

Click to view available options
Quantity:
2.5 L
4 L
Grade Hypergrade
Packaging Glass bottle
Boiling Point 100°C at 1013 hPa
Quantity 4 L