Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water 85% and Acetonitrile 15%, LabReady™ Solvent Blend, for LC-MS, for HPLC, Burdick & Jackson, Honeywell
Click to view available options
Quantity:
4 x 4 L
Description
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell's high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Specifications
CAS Max % | 87.90, 12.10 |
Grade | LC-MS |
Density | 0.9762 g/cm3 |
Boiling Point | 75°C |
Quantity | 4 x 4 L |
Flash Point | 23.3°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Safety and Handling
H370+H319+H226
P280+P210+P233+P240+P241+P242+P243+P260+P264+P270
Danger
Recommended Storage : Room Temperature